MilliporeSigma
  • Competitive amino acid transport between L-tryptophan and other amino acids in Schizophyllum commune.
Competitive amino acid transport between L-tryptophan and other amino acids in Schizophyllum commune.
Journal of bioscience and bioengineering (2005-10-20)
H Kinoshita, Y Maki, R Nakai, K Sen, H Shibai
ABSTRACT
In our study on nutritional requirement for the hyphal growth of Schizophyllum commune, we found that a Trp- mutant could not grow in the L-Trp-supplied medium in the presence of L-Ser. Further growth studies showed that not only L-Ser but also as many as 11 kinds of amino acid including L-Ala, L-Arg, L-Asn, L-His, L-Leu, L-Met, L-Phe, L-Ser, L-Thr, L-Tyr and L-Val inhibited the growth of the Trp- mutant in the L-Trp-supplied medium. However, these amino acids did not inhibit the growth of a Trp+ strain. The inhibition of growth of Trp+ strain induced by a Trp analogue of 5-fluoro-DL-tryptophan (5FT), which was usually recovered by L-Trp, was rescued by the same amino acids mentioned above. The exceptions were Gly and L-Ile, which also recovered the growth inhibition induced by 5FT. These results indicate that the permease responsible for the Trp transport in S. commune might also be active to other amino acids. However, it is considered that the permease shows high affinity to L-Trp and low affinity to other amino acids. As a result, the transport of L-Trp and 5FT may be counteracted by other amino acids.
Sigma-Aldrich
5-Fluoro-DL-tryptophan, powder or crystals