MilliporeSigma
  • Phosphaisocoumarins as a new class of potent inhibitors for pancreatic cholesterol esterase.
Phosphaisocoumarins as a new class of potent inhibitors for pancreatic cholesterol esterase.
European journal of medicinal chemistry (2010-02-13)
Baojian Li, Binhua Zhou, Hailiang Lu, Lin Ma, Ai-Yun Peng
ABSTRACT
Due to the importance of pancreatic cholesterol esterase (CEase) as a potential target in atherosclerosis and for the development of hypocholesterolemic agents, there are increasing interests in designing and synthesizing CEase inhibitors. In the present study, we prepared forty-five isocoumarin phosphorus analogues (i.e., phosphaisocoumarins) and investigated the inhibition of these compounds on the CEase. The results showed that some phosphaisocoumarins could act as potent inhibitors of CEase. The most potent inhibitors, compounds 9d, 10a and 12e give IC50 values of 4.8 microM, 2.3 microM and 1.9 microM, respectively. The inhibition mechanism and kinetic characterization studies indicate that they are reversible competitive inhibitors.
Sigma-Aldrich
Cholesterol Esterase from porcine pancreas, lyophilized, powder, white, ~35 U/mg