MilliporeSigma
Does Citrulline Have Protective Effects on Liver Injury in Septic Rats?
BioMed research international (2016-05-20)
Bin Cai, Yu-Long Luo, Shi-Jun Wang, Wei-Yuan Wei, Xue-Hui Zhang, Wei Huang, Tong Li, Meng Zhang, Nan Wu, Gopaul Roodrajeetsing, Sen Zhang
ABSTRACT
Citrulline (Cit) supplementation was proposed to serve as a therapeutic intervention to restore arginine (Arg) concentrations and improve related functions in sepsis. This study explored whether citrulline had positive effects on liver injury and cytokine release in the early stages of sepsis. The cecal ligation and puncture (CLP) model was utilized in our study. Rats were divided into four groups: normal, Cit, CLP, and CLP+Cit. The CLP group and CLP+Cit group were separated into 6-, 12-, and 24-hour groups, according to the time points of sacrifice after surgery. Intragastric administration of L-citrulline was applied to rats in Cit and CLP+Cit groups before surgery. Serum AST and ALT levels and levels of MDA, SOD, NO, and iNOS in the liver tissues were evaluated. Plasma concentrations of Cit and Arg were assessed using HPLC-MS/MS. Serum concentrations of cytokines and chemokines were calculated by Luminex. Results showed SOD activities of CLP+Cit groups were significantly higher than that of CLP groups, contrasting with the MDA and NO levels which were significantly lower in CLP+Cit groups than in CLP groups. In addition, plasma concentrations of TNF-α, IL-6, and IL-1β were significantly lower in the CLP+Cit 6-hour group than in the CLP 6-hour group.
Product Description
Millipore
MILLIPLEX® Rat Cytokine/Chemokine Magnetic Bead Panel - Premixed 27 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in rat serum, plasma and cell culture samples.