MilliporeSigma
  • Design, synthesis and dopamine D4 receptor binding activities of new N-heteroaromatic 5/6-ring Mannich bases.
Design, synthesis and dopamine D4 receptor binding activities of new N-heteroaromatic 5/6-ring Mannich bases.
Bioorganic & medicinal chemistry (2009-06-02)
Sabine Linz, Jörg Müller, Harald Hübner, Peter Gmeiner, Reinhard Troschütz
ABSTRACT
A series of phenylpiperazine-methyl-substituted 1H-pyrrolo[2,3-c]pyridine, imidazo[1,2-c]-, pyrrolo[2,3-d]- and pyrrolo[3,2-d]pyrimidines were prepared as selective dopamine D4-ligands. The pyrrolo[2,3-d]pyrimidine derivatives 12d (K(i)=1,9 nM) and 34 d (K(i)=2,4 nM) as well as the pyrrolo[3,2-d]pyrimidine Mannich base 49f (K(i)=2,8 nM) showed high dopamine D4 receptor activity superior to the atypical antipsychotic agent clozapine.
Sigma-Aldrich
1-Phenylpiperazine, ≥97.0%