MilliporeSigma
  • Role of the viral hemagglutinin in the anti-influenza virus activity of newly synthesized polycyclic amine compounds.
Role of the viral hemagglutinin in the anti-influenza virus activity of newly synthesized polycyclic amine compounds.
Antiviral research (2013-06-27)
Eva Torres, María D Duque, Evelien Vanderlinden, Chunlong Ma, Lawrence H Pinto, Pelayo Camps, Mathy Froeyen, Santiago Vázquez, Lieve Naesens
ABSTRACT
We here report on the synthesis of new series of polycyclic amines initially designed as ring-rearranged analogs of amantadine and featuring pentacyclo, hexacyclo, and octacyclo rings. A secondary amine, 3-azahexacyclo[7.6.0.0¹,⁵.0⁵,¹².0⁶,¹⁰.0¹¹,¹⁵]pentadeca-7,13-diene, 3, effectively inhibited A/M2 proton channel function, and, moreover, possessed dual activity against an A/H3N2 virus carrying a wild-type A/M2 proton channel, as well as an amantadine-resistant A/H1N1 virus. Among the polycyclic amines that did not inhibit influenza A/M2 proton channel function, several showed low-micromolar activity against tested A/H1N1 strains (in particular, the A/PR/8/34 strain), but not A/H3N2 influenza viruses. A/PR/8/34 mutants selected for resistance to these compounds possessed mutations in the viral hemagglutinin that markedly increased the hemolysis pH. Our data suggest that A/H1N1 viruses such as the A/PR/8/34 strain are particularly sensitive to a subtle increase in the endosomal pH, as caused by the polycyclic amine compounds.
Sigma-Aldrich
Anti-KDR/Flk-1/VEGFR2 Antibody, clone CH-11, clone CH-11, Upstate®, from mouse