MilliporeSigma
  • Role and prognostic significance of the Ki-67 index in non-Hodgkin's lymphoma.
Role and prognostic significance of the Ki-67 index in non-Hodgkin's lymphoma.
American journal of hematology (2009-04-23)
Adi Broyde, Olga Boycov, Yulia Strenov, Elimelech Okon, Ofer Shpilberg, Osnat Bairey
ABSTRACT
Expression of Ki-67, a nuclear antigen protein present in all cycling cells, is used to determine the growth fraction of tumors. The aim of this study was to evaluate the role and prognostic significance of the Ki-67 proliferation index (PI) in non-Hodgkin's lymphoma. Ki-67 was assayed immunohistochemically in tissue samples of 319 patients with newly-diagnosed non-Hodgkin's lymphoma. In 268 patients, the Ki-67 PI was correlated with clinical course and outcome. The mean Ki-67 PI ranged from 26.6% in indolent lymphomas to 97.6% in very aggressive lymphomas (P < 0.001). The index was <45% in 82.8% of indolent lymphomas and >45% in 85% of aggressive lymphomas (AUC = 0.877, P < 0.001). In patients with diffuse large B-cell lymphoma (n = 141), a Ki-67 PI of 70% was found to significantly discriminate patients with good or bad prognosis (AUC = 0.65, P = 0.004). Three-year survival was 75% +/- 5.6% in patients with a low Ki-67 index compared with 55.9% +/- 6% in patients with a high index (P = 0.015). In patients with a low IPI (<or=2), 3-year survival was 94% +/- 4% in those with a Ki-67 index <or=70% and 64% +/- 8.1% in those with a higher index (P = 0.002); in patients with bulky disease (>10 cm), the corresponding 3-year survival by Ki-67 index was 100% and 25% +/- 12% (P = 0.012). Our results suggest that the mean Ki-67 PI differs by type of lymphoma. A cut-off value of 45% can help differentiate indolent from aggressive disease. In diffuse large B-cell lymphoma, a cut-off value of 70% can distinguish patients with a good and bad prognosis when combined with other prognostic factors of low IPI score and bulky disease.
Sigma-Aldrich
Ki-67 (MRQ-64) Rabbit Monoclonal Antibody, clone MRQ-64, unconjugated, Cell Marque