MilliporeSigma
  • Synergistic effects of pazopanib and hyperthermia against uterine leiomyosarcoma growth mediated by downregulation of histone acetyltransferase 1.
Synergistic effects of pazopanib and hyperthermia against uterine leiomyosarcoma growth mediated by downregulation of histone acetyltransferase 1.
Journal of molecular medicine (Berlin, Germany) (2020-07-09)
Chiao-Yun Lin, Angel Chao, Ren-Chin Wu, Li-Yu Lee, Shir-Hwa Ueng, Chia-Lung Tsai, Yun-Shien Lee, Meng-Ting Peng, Lan-Yan Yang, Huei-Jean Huang, Hsin-Shih Wang, Chyong-Huey Lai
ABSTRACT
Pazopanib-a multitargeted tyrosine kinase inhibitor with prominent antiangiogenic effects-has shown promise in the treatment of soft-tissue sarcomas. Hyperthermia has been also applied as an adjunctive treatment to chemotherapy for these malignancies. Here, we show that pazopanib and hyperthermia act synergistically in inhibiting uterine leiomyosarcoma (LMS) cell growth. Compared with either treatment alone, the combination of pazopanib and hyperthermia exerted the highest antitumor activity in a xenograft model. Mechanistically, we found that combined treatment with pazopanib and hyperthermia inhibited histone acetyltransferase 1 (HAT1) expression in LMS cells. The Clock element on the HAT1 promoter was critical for pazopanib- and hyperthermia-induced HAT1 downregulation. Inhibition of HAT1-either by pazopanib and hyperthermia or through HAT1 silencing-was mediated by suppression of Clock. Accordingly, Clock protein reconstitution rescued both HAT1 levels and HAT1-mediated histone acetylation. Immunohistochemistry revealed a higher expression of HAT1 in uterine LMS than in leiomyomas (p = 0.007), with high HAT1 expression levels being associated with poor clinical outcomes (p = 0.007). We conclude that pazopanib and hyperthermia exert synergistic effects against LMS growth by inhibiting HAT1. Further preclinical studies on HAT1 as a potential drug target in uterine LMS are warranted, especially in combination with hyperthermia. KEY MESSAGES: Pazopanib and hyperthermia inhibit the growth of leiomyosarcoma. Their combined use inhibits HAT1 expression in leiomyosarcoma cells. The promoter Clock element is required for HAT1 downregulation. HAT1 expression is higher in leiomyosarcoma than in leiomyomas. An increased HAT1 expression is associated with poor clinical outcomes.
Sigma-Aldrich
Pazopanib hydrochloride, ≥98% (HPLC)