MilliporeSigma
  • Efficient transfer of cloned DNA into human diploid cells: protoplast fusion in suspension.
Efficient transfer of cloned DNA into human diploid cells: protoplast fusion in suspension.
Molecular and cellular biology (1984-11-01)
P Litzkas, K K Jha, H L Ozer
ABSTRACT
A method for fusion of protoplasts bearing amplified plasmids and human diploid fibroblasts or other cell types in suspension is described. Transient expression of plasmid-encoded proteins occurs in up to 50% of the human cells, as demonstrated for simian virus 40 T antigen by immunofluorescence and the Escherichia coli xanthine-guanine phosphoribosyl transferase by autoradiography. In contrast, frequencies of stable transformants were similar to those obtained by the CaPO4 coprecipitation technique. However, experiments with both methods involving the recombinant pRSVneo (in which the Rous sarcoma virus long terminal repeat regulates expression of the antibiotic-inactivating aminoglycoside phosphotransferase) revealed a much higher frequency of colonies in G418 selective medium with constructions in which the early region of simian virus 40 DNA was present as well. We propose a role for the simian virus 40 T antigen in enhancing stable transformation in this system.
Sigma-Aldrich
AC16 Human Cardiomyocyte Cell Line, AC16 Human Cardiomyocytes can be serially passaged and can differentiate when cultured in mitogen-free medium. The cells may be used to study developmental regulation of cardiomyocytes.