MilliporeSigma
  • 3, 3', 5-Triiodo-L-thyronine affects polarity proteins of bovine Sertoli cells via WT1/non-canonical Wnt signaling pathway.
3, 3', 5-Triiodo-L-thyronine affects polarity proteins of bovine Sertoli cells via WT1/non-canonical Wnt signaling pathway.
Theriogenology (2020-03-04)
Xue Wang, S O Adeniran, Ziming Wang, Xiaoyu Li, Fushuo Huang, Mingjun Ma, Zhongfeng Xu, Peng Zheng, Guixue Zhang
ABSTRACT
To determine the role of 3, 3', 5-triiodo-L thyroxine (T3) in the differentiation of Sertoli cells (SCs) and the factors influencing maturity via the Wilms' tumor 1 (WT1)/non-canonical Wnt signaling pathway, high purity SCs were isolated from newborn calves' testes and cultured in vitro. The SCs were stimulated with T3, and co-treated with short interference (si) RNA to knockdown endogenous WT1 and non-canonical Wnt signalling inhibitor Wnt-c59. Our results suggested that the addition of different concentrations (0, 25, 50, and 100 nM) of T3 in the culture medium changed the expression of KRT-18 (SCs immature marker) and accelerated the differentiation of SCs. T3 (100 nM) treatment induced up-regulated expression of WT1 over time (p < 0.05), while the expression of polarity proteins (Par3, Par6b, and E-cadherin) and Wnt4 were affected to varying degrees (p < 0.05). SCs were treated simultaneously with T3 + Wnt-c59 and T3 + WT1 siRNA, and the results showed that T3 could affect the expression of polarity proteins via WT1/non-canonical Wnt signaling pathway. These data put together indicate that T3 plays a dependent role in the induction of bovine SCs differentiation via WT1/non-canonical Wnt signaling pathway in vitro. This study proposes for the first time that WT1 is a major target for T3.
Sigma-Aldrich
MISSION® esiRNA, targeting human WT1