MilliporeSigma
  • In vitro alkylation of calf thymus DNA by acrylonitrile. Isolation of cyanoethyl-adducts of guanine and thymine and carboxyethyl-adducts of adenine and cytosine.
In vitro alkylation of calf thymus DNA by acrylonitrile. Isolation of cyanoethyl-adducts of guanine and thymine and carboxyethyl-adducts of adenine and cytosine.
Chemico-biological interactions (1984-09-15)
J J Solomon, I L Cote, M Wortman, K Decker, A Segal
ABSTRACT
Reaction of the rodent carcinogen acrylonitrile (AN) at pH 5.0 and/or pH 7.0 for 10 and/or 40 days with 2'-deoxyadenosine (dAdo), 2'-deoxycytidine (dCyd), 2'-deoxyguanosine (dGuo), 2'-deoxyinosine (dIno), N6-methyl-2'-deoxyadenosine (N6-Me-dAdo) and thymidine (dThd) resulted in the formation of cyanoethyl and carboxyethyl adducts. Adducts were not detected after 4 h. The adducts isolated were 1-(2-carboxyethyl)-dAdo (1-CE-dAdo), N6-CE-dAdo, 3-CE-dCyd, 7-(2-cyanoethyl)-Gua (7-CNE-Gua), 7,9-bis-CNE-Gua, imidazole ring-opened 7,9-bis-CNE-Gua, 1-CNE-dIno, 1-CE-N6-Me-dAdo and 3-CNE-dThd. Structures were assigned on the basis of UV spectra and electron impact (EI), chemical ionization (CI), desorption chemical ionization (DCI) and Californium-252 fission fragment ionization mass spectra. Evidence is presented which strongly suggests that N6-CE-dAdo was formed by Dimroth rearrangement of 1-CE-dAdo during the reaction between AN and dAdo. The carboxyethyl adducts resulted from initial cyanoethylation (by Michael addition) at a ring nitrogen adjacent to an exocyclic nitrogen atom followed by rapid hydrolysis of the nitrile moiety to a carboxylic acid. It was postulated that the facile hydrolysis is an autocatalyzed reaction resulting from the formation of a cyclic intermediate between nitrile carbon and exocyclic nitrogen. AN was reacted with calf thymus DNA (pH 7.0, 37 degrees C, 40 days) and the relative amounts of adducts isolated were 1-CE-Ade (26%), N6-CE-Ade (8%), 3-CE-Cyt (1%), 7-CNE-Gua (26%), 7,9-bis-CNE-Gua (4%), imidazole ring-opened 7,9-bis-CNE-Gua (19%) and 3-CNE-Thy (16%). Thus a carcinogen once adducted to a base in DNA was shown to be subsequently modified resulting in a mixed pattern of cyanoethylated and carboxyethylated AN-DNA adducts. Three of the adducts (1-CE-Ade, N6-CE-Ade and 3-CE-Cyt) were identical to adducts previously reported by us to be formed following in vitro reaction of the carcinogen beta-propiolactone (BPL) and calf thymus DNA. The results demonstrate that AN can directly alkylate DNA in vitro at a physiological pH and temperature.
Sigma-Aldrich
N6-Methyl-2′-deoxyadenosine