MilliporeSigma
  • Genistein protects against DSS-induced colitis by inhibiting NLRP3 inflammasome via TGR5-cAMP signaling.
Genistein protects against DSS-induced colitis by inhibiting NLRP3 inflammasome via TGR5-cAMP signaling.
International immunopharmacology (2019-03-23)
Yu Chen, Thi Ha Le, Qianming Du, Zheng Zhao, Yunxin Liu, Jianjun Zou, Weiwei Hua, Chao Liu, Yubing Zhu
ABSTRACT
NLRP3 inflammasome has been reported to be associated with inflammatory bowel disease including colitis due to its potential ability to induce IL-1β secretion. Emerging studies have demonstrated that Genistein, a major isoflavone, has potential anti-inflammatory effects in murine model colitis. However, its anti-inflammatory mechanism remains unclear. The effects of Genistein in dextran sulfate sodium (DSS)-induced murine colitis via targeting NLRP3 inflammasome was investigated in this study. Also, the mechanisms of protective action of Genistein in DSS-induced colitis may relate to TGR5 signaling. Genistein treatment not only remarkably attenuated loss of body weight and shortening of colon length but also significantly reduced inflammatory cells infiltration and pro-inflammatory mediator production in serum and colon. Moreover, Genistein treatment down-regulated production of caspase-1 and IL-1β and increased intracellular cAMP level, which were similar to the treatment for INT-777, a semi-synthetic TGR5 agonist, in phorbol myristate acetate (PMA)-differentiated monocytic THP-1 cells and U937 cells. These protective effects of Genistein might be attributed by ubiquination of NLRP3 which was induced due to interaction of cAMP with NLRP3. Furthermore, the effects of Genistein on NLRP3 inflammasome disappeared in TGR5-silenced U937 cells. In conclusion, our study unveils that Genistein was able to inhibit NLRP3 inflammasome via TGR5-cAMP signaling in macrophages. It therefore might be a potential effective drug for inflammatory bowel diseases.