MilliporeSigma
  • Pivmecillinam for Uncomplicated Lower Urinary Tract Infections Caused by Staphylococcus saprophyticus-Cumulative Observational Data from Four Recent Clinical Studies.
Pivmecillinam for Uncomplicated Lower Urinary Tract Infections Caused by Staphylococcus saprophyticus-Cumulative Observational Data from Four Recent Clinical Studies.
Antibiotics (Basel, Switzerland) (2019-05-10)
Filip Jansåker, Marianne Bollestad, Ingvild Vik, Morten Lindbæk, Lars Bjerrum, Niels Frimodt-Møller, Jenny Dahl Knudsen
ABSTRACT
Objectives: To investigate pivmecillinam´s efficacy in uncomplicated lower urinary tract infection (UTI) caused by Staphylococcus saprophyticus-considered non-susceptible to mecillinam. Methods: Participants with confirmed UTIs caused by S. saprophyticus from four randomized controlled trials, where pivmecillinam was empirically given to females with symptoms of UTIs. The primary outcome was defined as a cumulative clinical effect-symptom resolution during the first eight days of therapy, without a recurrence of UTI symptoms in the long-term follow-up (approximately four weeks). Secondary outcomes included the bacteriological effect-elimination of the causative agent, with or without new uropathogenic bacteria present in the first control urine sample. Significant bacteriuria was defined as ≥103 bacteria/mL. The antibiotic susceptibility testing was done by disc diffusion methodology, according to the European Committee on Antimicrobial Susceptibility Testing (EUCAST). Results: Seventy-four females (18-55 years) were empirically treated with pivmecillinam for UTIs caused by S. saphrophyticus (mean age 25 years; standard deviation (SD) 5.8). The cumulative clinical effect was 53/74 (72%), and the bacteriological effect was 51/59 (86%). Conclusion: Pivmecillinam showed a high clinical and bacteriological effect in UTIs caused by S. saprophyticus in these four clinical trials. The characterization of non-susceptibility for mecillinam regarding the treatment of UTIs caused by this common pathogen may need to be revised.