MilliporeSigma
  • Deposition and Source Identification of Nitrogen Heterocyclic Polycyclic Aromatic Compounds in Snow, Sediment, and Air Samples from the Athabasca Oil Sands Region.
Deposition and Source Identification of Nitrogen Heterocyclic Polycyclic Aromatic Compounds in Snow, Sediment, and Air Samples from the Athabasca Oil Sands Region.
Environmental science & technology (2019-02-12)
Leah Chibwe, Carlos A Manzano, Derek Muir, Beau Atkinson, Jane L Kirk, Christopher H Marvin, Xiaowa Wang, Camilla Teixeira, Dayue Shang, Tom Harner, Amila O De Silva
ABSTRACT
Polycyclic aromatic compounds (PACs) can have multiple sources in the Athabasca Oil Sands Region (AOSR). The current study was designed to identify and explore the potential of nitrogen heterocyclic PACs (NPACs) as source indicators in snowpack, lake sediment and passive air samples from the AOSR during 2014-2015. Source samples including petroleum coke (petcoke), haul road dust, and unprocessed oil sands were also analyzed. Samples were analyzed using comprehensive two-dimensional gas chromatography time-of-flight mass spectrometry, and liquid chromatography-high resolution Orbitrap mass spectrometry. Over 200 NPACs were identified and classified into at least 24 isomer groups, including alkylated carbazoles, benzocarbazoles, and indenoquinolines. Levels of NPACs in environmental samples decreased with distance from the main developments and with increasing depth in lake sediments but were detected within 50 km from the major developments. The composition profiles of several NPAC isomer classes, such as dimethylcarbazoles, showed that petcoke had a distinct distribution of NPACs compared to the haul road dust and unprocessed oil sands ores and was the most similar source material to near-field environmental samples. These results suggest that petcoke is a major contributing source for the identified NPACs and that these compounds have the potential to be used as source indicators for future research in the AOSR.