MilliporeSigma
  • Dynamic agent of an injectable and self-healing drug-loaded hydrogel for embolization therapy.
Dynamic agent of an injectable and self-healing drug-loaded hydrogel for embolization therapy.
Colloids and surfaces. B, Biointerfaces (2018-09-17)
Xiaqing Zhou, Yongsan Li, Shuang Chen, Ya-Nan Fu, Shihui Wang, Guofeng Li, Lei Tao, Yen Wei, Xing Wang, Jun F Liang
ABSTRACT
Embolic agents are crucial for trans-catheter arterial embolization (TAE) in the treatment of various unresectable malignant tumors. Although solid particles, liquid oils, and some polymeric hydrogels have proved their capacities for embolic therapies, the low efficiency, time sensitivity, and cytotoxicity are still considered as challenges. In this study, we developed a three-component dynamic self-healing hydrogel to overcome these limitations. With the help of the Schiff-base bonding, both glycol-chitosan and carbazochrome, containing amine groups, react with dibenzaldehyde-terminated poly(ethylene-glycol) (DF-PEG), forming the dynamic self-healing hydrogels under a mild condition within 200 s. 1H NMR and rheology test were used to characterize the Schiff-base formation and mechanical strength. Controlled-release of carbazochrome from different gelator concentrations of DF-PEG was also studied. Furthermore, in vivo evaluation of the embolization on rats showed the superior embolic effects of the injectable and self-healing hydrogel. Therefore, this new dynamic agent demonstrated the potential for application as a simple, inexpensive, and tunable embolic agent for cancer treatment and drug delivery system.