MilliporeSigma
  • Synthesis and Biological Activity of Some Benzochromenoquinolinones: Tacrine Analogs as Potent Anti-Alzheimer's Agents.
Synthesis and Biological Activity of Some Benzochromenoquinolinones: Tacrine Analogs as Potent Anti-Alzheimer's Agents.
Chemistry & biodiversity (2019-02-06)
Mohammad Mahdavi, Roshanak Hariri, Seyedeh Sara Mirfazli, Hania Lotfian, Arezoo Rastergari, Omidreza Firuzi, Najmeh Edraki, Bagher Larijani, Tahmineh Akbarzadeh, Mina Saeedi
ABSTRACT
Alzheimer's disease (AD) is a well-known neurodegenerative disorder affecting millions of old people worldwide and the corresponding epidemiological data emphasize the importance of the disease. As AD is a multifactorial illness, various single target directed drugs that have reached clinical trials have failed. Therefore, various factors associated with outset of AD have been considered in targeted drug discovery. In this work, various benzochromenoquinolinones were synthesized and evaluated for their cholinesterase and BACE1 inhibitory activities as well as neuroprotective and metal-chelating properties. Among the synthesized compounds, 14-amino-13-(3-nitrophenyl)-2,3,4,13-tetrahydro-1H-benzo[6,7]chromeno[2,3-b]quinoline-7,12-dione (6m) depicted the best inhibitory activity toward acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) with IC50 s of 0.86 and 6.03 μm, respectively. Also, the compound could inhibit β-secretase 1 (BACE1) with IC50 =19.60 μm and showed metal chelating ability toward Cu2+ , Fe2+ , and Zn2+ . In addition, docking study demonstrated desirable interactions of compound 6m with amino acid residues characterizing AChE, BChE, and BACE1.