MilliporeSigma
  • MicroRNA-497-5p inhibits proliferation and invasion of non-small cell lung cancer by regulating FGF2.
MicroRNA-497-5p inhibits proliferation and invasion of non-small cell lung cancer by regulating FGF2.
Oncology letters (2019-03-15)
Xiaori Huang, Lei Wang, Wei Liu, Fei Li
ABSTRACT
Increasing number of microRNAs (miRNAs) have been reported to play an important role in the development and progression of non-small cell lung cancer (NSCLC). In particular, microRNA-497-5p (miR-497-5p) has been proposed as a tumor suppressor miRNA in human cancers. However, the role of miR-497-5p and its potential molecular mechanism associated with NSCLC are less studied. Therefore, the role of miR-497-5p in the pathogenesis of NSCLC was investigated. In the present study, the expression of miR-497-5p was significantly downregulated in NSCLC. Moreover, overexpression of miR-497-5p inhibited the proliferation and invasion of NSCLC cells by suppressing FGF2. In addition, FGF2 was a downstream target of miR-497-5p in NSCLC. FGF2 was upregulated in NSCLC promoting cell proliferation and invasion. Overexpression of FGF2 impaired the inhibitory effect of miR-497-5p in NSCLC. Taken together, these results demonstrate that miR-497-5p is a tumor suppressor miRNA and demonstrate its potential for future use in the treatment of human NSCLC.