MilliporeSigma
  • AKT Inhibition Modulates H3K4 Demethylase Levels in PTEN-Null Prostate Cancer.
AKT Inhibition Modulates H3K4 Demethylase Levels in PTEN-Null Prostate Cancer.
Molecular cancer therapeutics (2018-11-18)
Mohammad Imran Khan, Abid Hamid, Suvasmita Rath, Bushra Ateeq, Qateeb Khan, Imtiaz A Siddiqui, Vaqar Mustafa Adhami, Hani Choudhry, Mazin A Zamzami, Hasan Mukhtar
ABSTRACT
Hyperactivated AKT kinase due to loss of its negative regulator PTEN influences many aspects of cancer biology, including chromatin. AKT primarily regulates acetyl-CoA production and phosphorylates many histone-modulating enzymes, resulting in their activation or inhibition. Therefore, understanding the therapeutic impact of AKT inhibition on chromatin-related events is essential. Here, we report that AKT inhibition in prostate-specific PTEN knockout mice significantly induces di- and trimethylation of H3K4 with concomitant reduction in H3K9 acetylation. Mechanistically, we observed that AKT inhibition reduces expression of the H3K4 methylation-specific histone demethylases KDM5 family, especially KDM5B expression at transcriptional levels. Furthermore, we observed that AKT negatively regulates miR-137 levels, which transcriptionally represses KDM5B expression. Overexpression of miR-137 significantly reduced KDM5B and increased H3K4 methylation levels but failed to change AKT phosphorylation. Overall, we observed that AKT transcriptionally regulates KDM5B mainly via repression of miR-137. Our data identify a mechanism by which AKT kinase modulates the prostate cancer epigenome through regulating H3K4 methylation. Additional studies on AKT inhibition-mediated induction of H3K4 methylation will help in designing strategies to enhance the therapeutic efficacy of PI3K/AKT inhibitors.