MilliporeSigma
  • Effects of S-metolachlor and its degradation product metolachlor OA on marbled crayfish (Procambarus virginalis).
Effects of S-metolachlor and its degradation product metolachlor OA on marbled crayfish (Procambarus virginalis).
Chemosphere (2019-03-09)
Alzbeta Stara, Jan Kubec, Eliska Zuskova, Milos Buric, Caterina Faggio, Antonin Kouba, Josef Velisek
ABSTRACT
Increasing production of energy crops in Europe, mainly maize and rapeseed, has altered patterns of pesticide use in recent decades. The long-term effects of S-metolachlor (S-M) and of its metabolite metolachlor OA (M-OA) at the environmentally relevant concentration of 4.2 μg L-1 and at 42 μg L-1 (ten-fold concentration) on marbled crayfish (Procambarus virginalis) were evaluated in a 28-day exposure and after a subsequent 28-day recovery period. Indicators assessed were behaviour; biochemical haemolymph profile; oxidative and antioxidant parameters of gill, hepatopancreas, and muscle; and histology of hepatopancreas and gill. Results showed biochemical haemolymph profile (lactate, alanine aminotransferase, aspartate aminotransferase, inorganic phosphate), lipid peroxidation in hepatopancreas, and antioxidant parameters (catalase, reduced glutathione, glutathione S-transferase) of hepatopancreas and gill of crayfish exposed to S-M and M-OA to significantly differ from controls (P < 0.01). Antioxidant biomarker levels remained different from controls after a 28-day recovery period. Differences in behaviour including speed of movement and velocity, and histopathological damage to gill and hepatopancreas were associated with S-M and M-OA exposure and persisted after 28 days in S-M- and M-OA-free water. Results suggest harmful effects of low concentrations of S-M and its metabolite M-OA on non-target organisms and provide information for assessing their effects at environmentally relevant concentrations.