MilliporeSigma
  • Pilot, Open, Randomized, Prospective Trial for Normothermic Machine Perfusion Evaluation in Liver Transplantation From Older Donors.
Pilot, Open, Randomized, Prospective Trial for Normothermic Machine Perfusion Evaluation in Liver Transplantation From Older Donors.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society (2018-10-27)
Davide Ghinolfi, Erion Rreka, Vincenzo De Tata, Maria Franzini, Daniele Pezzati, Vanna Fierabracci, Matilde Masini, Andrea Cacciatoinsilla, Maria Lucia Bindi, Lorella Marselli, Valentina Mazzotti, Riccardo Morganti, Piero Marchetti, Giandomenico Biancofiore, Daniela Campani, Aldo Paolicchi, Paolo De Simone
ABSTRACT
Ex situ normothermic machine perfusion (NMP) might minimize ischemia/reperfusion injury (IRI) of liver grafts. In this study, 20 primary liver transplantation recipients of older grafts (≥70 years) were randomized 1:1 to NMP or cold storage (CS) groups. The primary study endpoint was to evaluate graft and patient survival at 6 months posttransplantation. The secondary endpoint was to evaluate liver and bile duct biopsies; IRI by means of peak transaminases within 7 days after surgery; and incidence of biliary complications at month 6. Liver and bile duct biopsies were collected at bench surgery, end of ex situ NMP, and end of transplant surgery. Interleukin (IL) 6, IL10, and tumor necrosis factor α (TNF-α) perfusate concentrations were tested during NMP. All grafts were successfully transplanted. Median (interquartile range) posttransplant aspartate aminotransferase peak was 709 (371-1575) IU/L for NMP and 574 (377-1162) IU/L for CS (P = 0.597). There was 1 hepatic artery thrombosis in the NMP group and 1 death in the CS group. In NMP, we observed high TNF-α perfusate levels, and these were inversely correlated with lactate (P < 0.001). Electron microscopy showed decreased mitochondrial volume density and steatosis and an increased volume density of autophagic vacuoles at the end of transplantation in NMP versus CS patients (P < 0.001). Use of NMP with older liver grafts is associated with histological evidence of reduced IRI, although the clinical benefit remains to be demonstrated.