MilliporeSigma
Aminothiazoles as γ-secretase modulators.
Bioorganic & medicinal chemistry letters (2011-09-20)
Thomas Lübbers, Alexander Flohr, Synese Jolidon, Pascale David-Pierson, Helmut Jacobsen, Laurence Ozmen, Karlheinz Baumann
ABSTRACT
We herein report the discovery of a new γ-secretase modulator class with an aminothiazole core starting from a HTS hit (3). Synthesis and SAR of this series are discussed. These novel compounds demonstrate moderate to good in vitro potency in inhibiting amyloid beta (Aβ) peptide production. Overall γ-secretase is not inhibited but the formation of the aggregating, toxic Aβ42 peptide is shifted to smaller non-aggregating Aβ peptides. Compound 15 reduced brain Aβ42 in vivo in APPSwe transgenic mice at 30mg/kg p.o.
Sigma-Aldrich
1-Methylpyrazole-4-boronic acid pinacol ester, 95%