MilliporeSigma
  • DNA containing side chains with terminal triple bonds: Base-pair stability and functionalization of alkynylated pyrimidines and 7-deazapurines.
DNA containing side chains with terminal triple bonds: Base-pair stability and functionalization of alkynylated pyrimidines and 7-deazapurines.
Chemistry & biodiversity (2006-12-29)
Frank Seela, Venkata Ramana Sirivolu
ABSTRACT
The synthesis of a series of oligonucleotides containing 5-substituted pyrimidines as well as 7-substituted 7-deazapurines bearing diyne groups with terminal triple bonds is reported. The modified nucleosides were prepared from the corresponding iodo nucleosides and diynes by the Sonogashira cross-coupling reaction. They were converted into phosphoramidites and employed in solid-phase synthesis of oligonucleotides. The effect of the diyne modifications on the duplex stability was investigated. The modified nucleosides were used for further functionalization using the protocol of Huisgen-Sharpless [2+3] cycloaddition ('click chemistry').
Sigma-Aldrich
C8-Alkyne-dU-CEP