MilliporeSigma
  • The fructokinase from Rhizobium leguminosarum biovar trifolii belongs to group I fructokinase enzymes and is encoded separately from other carbohydrate metabolism enzymes.
The fructokinase from Rhizobium leguminosarum biovar trifolii belongs to group I fructokinase enzymes and is encoded separately from other carbohydrate metabolism enzymes.
Microbiology (Reading, England) (1996-02-01)
G J Fennington, T A Hughes
ABSTRACT
The Rhizobium leguminosarum bv. trifolii BAL fructokinase (frk) gene was isolated on a 2 center dot 4 kb BamHI fragment from the cosmid pLA72 by complementation analysis of the Tn5-induced frk mutant BAL79, and confirmed by hybridization analysis. The nucleotide sequence of the frk gene was found to contain an open reading frame consisting of 978 bp encoding 326 amino acids, which was then compared to known fructokinase sequences. The fructokinase gene was not contained in an operon and is encoded separately from other enzymes of carbohydrate metabolism. Its product is therefore assigned to the group I fructokinases. A putative promoter (TTGACA-N16-GTTGAT), ribosome-binding site and termination sequence were identified. The Frk protein contained several motifs conserved in other known fructokinase sequences, including an ATP-binding and a substrate-binding motif. The hydropathy plot derived from the frk gene sequence data revealed the fructokinase as a hydrophilic protein. The fructokinase protein was purified to electrophoretic homogeneity by a three-step method using chromatofocusing, affinity chromatography and gel filtration. Its purity was confirmed by SDS-PAGE and it was visualized as a single band by silver staining. The N-terminal amino acid sequence of the purified fructokinase confirmed the proposed open reading frame of the frk gene. The purified fructokinase had a molecular mass of 36 center dot 5 kDa, pl of 4 center dot 65, pH activity range of 6 center dot 0-9 center dot 0 (maximum activity at pH 8 center dot 0) and a Mg2+ requirement. It had a Km of 0 center dot 31 mM and a Vmax of 31 mumol fructose 6-phosphate (mg protein)-1 min-1 with fructose as substrate. The R. leguminosarum bv. trifolii BAL fructokinase was biochemically and molecularly similar to other bacterial fructokinases.
Sigma-Aldrich
Epoxy-activated-Agarose, lyophilized powder