MilliporeSigma
  • The protein SET regulates the inhibitory effect of p21(Cip1) on cyclin E-cyclin-dependent kinase 2 activity.
The protein SET regulates the inhibitory effect of p21(Cip1) on cyclin E-cyclin-dependent kinase 2 activity.
The Journal of biological chemistry (1999-11-07)
J M Estanyol, M Jaumot, O Casanovas, A Rodriguez-Vilarrupla, N Agell, O Bachs
ABSTRACT
The cyclin-dependent kinase (CDK) inhibitor p21(Cip1) has a dual role in the regulation of the cell cycle; it is an activator of cyclin D1-CDK4 complexes and an inhibitor of cyclins E/A-CDK2 activity. By affinity chromatography with p21(Cip1)-Sepharose 4B columns, we purified a 39-kDa protein, which was identified by microsequence analysis as the oncoprotein SET. Complexes containing SET and p21(Cip1) were detected in vivo by immunoprecipitation of Namalwa cell extracts using specific anti-p21(Cip1) antibodies. We found that SET bound directly to p21(Cip1) in vitro by the carboxyl-terminal region of p21(Cip1). SET had no direct effect on cyclin E/A-CDK2 activity, although it reversed the inhibition of cyclin E-CDK2, but not of cyclin A-CDK2, induced by p21(Cip1). This result is specific for p21(Cip1), since SET neither bound to p27(Kip1) nor reversed its inhibitory effect on cyclin E-CDK2 or cyclin A-CDK2. Thus, SET appears to be a modulator of p21(Cip1) inhibitory function. These results suggest that SET can regulate G(1)/S transition by modulating the activity of cyclin E-CDK2.
Sigma-Aldrich
Anti-p21WAF1 (Ab-1) Mouse mAb (EA10), liquid, clone EA10, Calbiochem®