MilliporeSigma
  • The fluid membrane determines mechanics of erythrocyte extracellular vesicles and is softened in hereditary spherocytosis.
The fluid membrane determines mechanics of erythrocyte extracellular vesicles and is softened in hereditary spherocytosis.
Nature communications (2018-11-25)
Daan Vorselen, Susan M van Dommelen, Raya Sorkin, Melissa C Piontek, Jürgen Schiller, Sander T Döpp, Sander A A Kooijmans, Brigitte A van Oirschot, Birgitta A Versluijs, Marc B Bierings, Richard van Wijk, Raymond M Schiffelers, Gijs J L Wuite, Wouter H Roos
ABSTRACT
Extracellular vesicles (EVs) are widely studied regarding their role in cell-to-cell communication and disease, as well as for applications as biomarkers or drug delivery vehicles. EVs contain membrane and intraluminal proteins, affecting their structure and thereby likely their functioning. Here, we use atomic force microscopy for mechanical characterization of erythrocyte, or red blood cell (RBC), EVs from healthy individuals and from patients with hereditary spherocytosis (HS) due to ankyrin deficiency. While these EVs are packed with proteins, their response to indentation resembles that of fluid liposomes lacking proteins. The bending modulus of RBC EVs of healthy donors is ~15 kbT, similar to the RBC membrane. Surprisingly, whereas RBCs become more rigid in HS, patient EVs have a significantly (~40%) lower bending modulus than donor EVs. These results shed light on the mechanism and effects of EV budding and might explain the reported increase in vesiculation of RBCs in HS patients.
Sigma-Aldrich
Monoclonal Anti-Band 3 antibody produced in mouse, clone BIII-136, ascites fluid