MilliporeSigma
  • Early changes in transient adenosine during cerebral ischemia and reperfusion injury.
Early changes in transient adenosine during cerebral ischemia and reperfusion injury.
PloS one (2018-05-26)
Mallikarjunarao Ganesana, B Jill Venton
ABSTRACT
Adenosine is an important neuromodulator in the central nervous system, and tissue adenosine levels increase during ischemic events, attenuating excitotoxic neuronal injury. Recently, our lab developed an electrochemical fast-scan cyclic voltammetry (FSCV) method that identified rapid, spontaneous changes in adenosine concentrations that last only about 3 seconds. Here, we investigated the effects of cerebral ischemia and reperfusion on the concentration and frequency of transient adenosine release in the caudate-putamen. In anesthetized rats, data were collected for four hours: two hours of normoxia, 30 min of cerebral ischemia induced by bilateral common carotid artery occlusion, and 90 min of reperfusion. Transient adenosine release was increased during the cerebral ischemia period and remained elevated during reperfusion. The total number of adenosine transients increased by 52% during cerebral ischemia and reperfusion compared to normoxia. The concentration of adenosine per event did not increase but the cumulative adenosine concentration during cerebral ischemia and reperfusion increased by 53% because of the higher frequency of events. Further, we evaluated the role of A2A antagonist, SCH442416, a putative neuroprotective agent to affect adenosine transients. SCH442416 significantly decreased the transient frequency during cerebral ischemia-reperfusion by 27% and the cumulative concentration by 31%. Our results demonstrate that this mode of rapid adenosine release increases during early cerebral ischemia-reperfusion injury. Rapid adenosine release could provide fast, local neuromodulation and neuroprotection during cerebral ischemia.
Sigma-Aldrich
SCH-442416, ≥98% (HPLC)