Skip to Content
MilliporeSigma
Vitamin K1 2,3-epoxide ≥98.0% (HPLC)

Vitamin K1 2,3-epoxide

Vitamin K1 2,3-epoxide ≥98.0% (HPLC)
Synonym(s):
(2,3-Epoxyphytyl)menaquinone, 1a,7a-Dihydro-1a-methyl-7a-(3,7,11,15-tetramethyl-2-hexadecenyl)naphth[2,3-b]oxirene-2,7-dione, 2,3-Epoxy-2,3-dihydro-2-methyl-3-phytyl-1,4-naphthoquinone, 2,3-Epoxyphylloquinone, Phylloquinone oxide
Empirical Formula (Hill Notation):
C31H46O3
CAS Number:
Molecular Weight:
466.70
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing