
Panicolin

Synonym(s):
Skullcapflavone I, 2′,5-Dihydroxy-7,8-dimethoxyflavone, 5-Hydroxy-2-(2-hydroxyphenyl)-7,8-dimethoxy-4H-1-benzopyran-4-one
Empirical Formula (Hill Notation):
C17H14O6
CAS Number:
Molecular Weight:
314.29
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1