Skip to Content
MilliporeSigma
vibrant-m

ORLISTAT RELATED COMPOUND D SOLUTION

vibrant-m
Synonym(s):
N-Formyl-L-leucine (3S,4R,6S)-3-hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl ester
Empirical Formula (Hill Notation):
C29H53NO5
CAS Number:
Molecular Weight:
495.73

Products

Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1