
Beflubutamid

Synonym(s):
2-[4-Fluoro-3-(trifluoromethyl)phenoxy]-N-(phenylmethyl)butanamide, N-Benzyl-2-(α,α,α,4-tetrafluoro-m-tolyloxy)butyramide
Empirical Formula (Hill Notation):
C18H17F4NO2
CAS Number:
Molecular Weight:
355.33
Products
Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1