Product Name
3-Isobutyl-1-methylxanthine ≥99%, BioUltra
Product Number
I7018
Product Brand
SIGMA
CAS Number
28822-58-4
Molecular Weight
222.24
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 98.5 %
SULFATED ASH≤ 0.1 %
ALUMINIUM (ICP)≤ 5 mg/kg
CALCIUM (ICP)≤ 50 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 50 mg/kg
POTASSIUM (ICP)≤ 500 mg/kg
MAGNESIUM (ICP)≤ 50 mg/kg
SODIUM (ICP)≤ 50 mg/kg
LEAD (ICP)≤ 10 mg/kg
ZINC (ICP)≤ 5 mg/kg
TOTAL PHOSPHORUS AS PO4 (ICP)≤ 50 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 500 mg/kg
ION CHROMATOGRAPHY (NH4+)≤ 500 mg/kg