Product Name
Bis(trimethylsilyl) acetylenedicarboxylate ≥98.0%
Product Number
01095
Product Brand
SIAL
CAS Number
76734-92-4
Molecular Weight
258.42
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow and Colorless to Light Green-Yellow |
APPEARANCE (FORM) | Liquid |
TITRATION (T) NAOH 0.1M | 98.0 - 102.0 % |
REFRACTIVE INDEX N20/D | 1.438 - 1.440 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |