Product Name
3-Bromophenyl isopropyl ether 97%
Product Number
515507
Product Brand
ALDRICH
CAS Number
131738-73-3
Molecular Weight
215.09
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
TITRATION96.5 - 103.5 %
TITRATION (METHOD)WITH AGNO3 AFTER OXYGEN COMBUSTION
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE