Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13269E

Centrisart® I centrifugal ultrafiltration unit

MWCO 100 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

material

polyethersulfone membrane
polystyrene housing
polystyrene outer tube

sterility

non-sterile

packaging

pack of 12 tubes

manufacturer/tradename

Sartorius 13269E

parameter

2000 × g max. force (fixed angle)
2500 × g max. force (swinging bucket)

filtration area

0.79 cm2

outer sample tube W × L

14 mm × 93 mm

tube size

~15 mL

working volume × concentrate volume

2.5 mL × 100

pore size

100 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

  • Centrisart® I centrifugal ultrafiltration unit is ideal for the separation of small-volume proteins from low molecular weight substances in biological samples.
  • The unit works in the direction opposite to centrifugal force which prevents premature filter blockage.
  • This versatile device is utilized in both fixed-angle or swinging-bucket rotors that hold standard 15 mL tubes.
  • The product is collected in the easily accessible floater insert tube.

Application

  • Protein removal from blood samples
  • Drug binding studies
  • Cleaning of liposomes (cell lysates)

Legal Information

Centrisart is a registered trademark of Sartorius Stedim Biotech GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service