Skip to Content
MilliporeSigma
All Photos(3)

Documents

HPA003203

Sigma-Aldrich

Anti-ZNF764 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger protein 764 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VYFCREEWGCLRPAQRALYRDVMRETYGHLSALGIGGNKPALISWVEEEAELWGPAAQDPEVAKCQTQTDPDSRNKKKERQREGTGALEKPDPVAAGSPGLKSPQAPSAGPPYGWEQLSKAPHRGRPSLCAHPPVPRADQRH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF764(92595)

General description

The zinc finger proteins (ZNFs) are made up of anti-parallel hairpin motif. These proteins contain an α-helix, two β-strands and a hairpin structure. The ZNFs are mainly involved in the recognition of DNA, folding and assembly of proteins, transcriptional activation, lipid binding, regulation of apoptosis, and packaging of RNA.

Immunogen

Zinc finger protein 764 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZNF764 (zinc finger protein 764) gene encodes a protein belonging to the kruppel C2H2-type zinc-finger protein family. It may function as a transcriptional regulator by interacting with a general nuclear hormone receptor coactivator, transcriptional intermediary factor 1. It is involved in glucocorticoid, androgen, and thyroid hormone action and its microdeletion leads to haploinsufficiency causing partial multiple hormone resistance.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83542

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoshige Kino et al.
The Journal of clinical endocrinology and metabolism, 97(8), E1557-E1566 (2012-05-12)
Nuclear hormone receptors exert their transcriptional effects through shared cofactor molecules; thus, defects in such intermediate proteins may be associated with multiple hormone resistance. Microdeletion of small chromosomal segments results in hereditary or sporadic diseases by affecting expression of residing
Identification and Analysis of Disease Target Network of Human MicroRNA and Predicting Promising Leads for ZNF439, a Potential Target for Breast Cancer.
Raghavan V and Manasa D B
International Journal of Bioscience, Biochemistry and Bioinformatics, 2, 358-362 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service