Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73310

Sigma-Aldrich

4-Nitrophenyl phenyl ether

purum, ≥98.0% (GC)

Synonym(s):

1-Nitro-4-phenoxybenzene, 4-Nitrodiphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OC6H5
CAS Number:
Molecular Weight:
215.20
Beilstein/REAXYS Number:
2051911
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

assay

≥98.0% (GC)

bp

320 °C (lit.)

mp

53-56 °C (lit.)
56-60 °C

SMILES string

[O-][N+](=O)c1ccc(Oc2ccccc2)cc1

InChI

1S/C12H9NO3/c14-13(15)10-6-8-12(9-7-10)16-11-4-2-1-3-5-11/h1-9H

InChI key

JDTMUJBWSGNMGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service