Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ST1676

Sigma-Aldrich

Anti-TMPRSS2 Mouse mAb (2F4)

liquid, clone 2F4, Calbiochem®

Synonym(s):

Anti-Transmembrane Protease Serine 2, Anti-Epitheliasin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F4, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TMPRSS2(7113)

General description

Purified mouse monoclonal antibody. Recognizes the TMPRSS2 protein.
Recognizes the TMPRSS2 protein.
This Anti-TMPRSS2 Mouse mAb (2F4) is validated for use in ELISA, Immunoblotting for the detection of TMPRSS2.

Immunogen

A recombinant polypeptide corresponding to amino acids of 383-493 human TMPRSS2, expressed as a GST fusion protein

Application

ELISA (see comments)

Immunoblotting (1-5 µg/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service