Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX0530

Supelco

Phenol Red

pH indicator, ACS, GR ACS

Synonym(s):

Phenol Red, Phenolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14O5S
CAS Number:
Molecular Weight:
354.38
MDL number:
UNSPSC Code:
12171502
EC Index Number:
205-609-7

Product Name

Phenol Red, Meets ACS Specifications GR ACS

form

powder

loss

≤1% loss on drying

color

red

visual transition interval

6.8-8.2, yellow to red

mp

>300 °C

solubility

0.77 g/L

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C19H14O5S/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19/h1-12,20-21H

Inchi Key

BELBBZDIHDAJOR-UHFFFAOYSA-N

Application

Meets ACS Specifications GR ACS

Analysis Note

Clarity of solutionTo pass testColorRedFormPowderLoss on drying1% maxVisual transition intervalTo pass testVisual transition, pH 6.8YellowVisual transition, pH 8.2Red

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service