Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PF048

Sigma-Aldrich

Heregulin, Human, Recombinant, E. coli

Synonym(s):

Neu Differentiation Marker, NDF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human heregulin expressed in E. coli. Heregulins have been shown to be mitogenic for Schwann cells and various epithelial cells. It is recommended that the protein be titrated for optimal results in individual systems.
Recombinant, human heregulin expressed in E. coli. Human recombinant heregulin whose DNA sequence encoding the EGF domain of heregulin α (amino acids 177-241) expressed in and purified from E. coli. The expressed protein consists of 65 amino acids with a predicted molecular weight of approximately 7 kDa.

Application

Proliferation Studies

Biochem/physiol Actions

EC₅₀ ~20-40 ng/ml as determined by a cell proliferation assay using breast carcinoma cell line SK-BR-3 or in a serum-free cell proliferation assay using the human cell line MCF7.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from PBS, 50 µg BSA/µg cytokine.

Reconstitution

Store lyophilized protein dessicated at -20°C. Lyophilized product should be reconstituted in sterile PBS containing at least 0.1% human serum albumin or bovine serum albumin to prepare a stock solution of no less than 50 µg/ml. Following initial thaw, aliquot and freeze (-20°C).

Other Notes

It is recommended that the protein be titrated for optimal results in individual systems.
Riese, D.J., et al. 1996. J. Biol. Chem.271, 20047.
Holmes, W.E., et al. 1992. Science256, 1205.
Karey, K.P., et al. 1988. Cancer Res.48, 4083.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maximilian Haertinger et al.
Cells, 9(1) (2020-01-16)
Recent studies showed a beneficial effect of adipose stem cell-derived extracellular vesicles (ADSC-EVs) on sciatic nerve repair, presumably through Schwann cell (SC) modulation. However, it has not yet been elucidated whether ADSC-EVs exert this supportive effect on SCs by extracellular
Rachel R Besser et al.
Biomaterials science, 8(2), 591-606 (2019-12-21)
We report a water-soluble and non-toxic method to incorporate additional extracellular matrix proteins into gelatin hydrogels, while obviating the use of chemical crosslinkers such as glutaraldehyde. Gelatin hydrogels were fabricated using a range of gelatin concentrations (4%-10%) that corresponded to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service