573131
STAT3 Inhibitor XIV, LLL12 - Calbiochem
The STAT3 Inhibitor XIV, LLL12 controls the biological activity of STAT3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
STAT3 Inhibitor XIV, LLL12 - Calbiochem, 5-Hydroxy-9,10-dioxo-9,10-dihydroanthracene-1-sulfonamide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C14H9NO5S
Molecular Weight:
303.29
UNSPSC Code:
12352200
Recommended Products
Quality Level
assay
≥98% (HPLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
orange
solubility
DMSO: 10 mg/mL
shipped in
ambient
storage temp.
2-8°C
General description
A cell-permeable hydroxyanthraquinone compound that binds directly to the phosphoryl Tyr705 binding site of the STAT3 monomer and acts as selective and reversible inhibitor of STAT3 signaling. Shown to block STAT3 phosphorylation and its subsequent nuclear localization and DNA binding, but has no effect on IL-4, interferon-α and interferon-γ-induced STAT1, 2, 4, 6 and pJAK2, PI3-Kinase, or ERK signaling. Does not affect the activity of over 21 other closely related protein kinases (IC50 >50 µM). Also shown to induce apoptosis in U266 and ARH-77 multiple myeloma cell lines (IC50 = 490 nM and 1.96 µM, respectively), and suppress hepatocellular carcinoma tumor growth in xenograft mouse model (5 mg/kg, i.v).
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Other Notes
Lin, L., et al. 2011. Cancer Res.71, 7226.
Lin, L., et al. 2011. Int. J. Cancerin press.
Ball, S., et al. PLoS One 6, e18820.
Liu, Y., et al. 2010. J. Biol. Chem. 285, 27429.
Lin, L., et al. 2010. Neoplasia12, 39.
Lin, L., et al. 2011. Int. J. Cancerin press.
Ball, S., et al. PLoS One 6, e18820.
Liu, Y., et al. 2010. J. Biol. Chem. 285, 27429.
Lin, L., et al. 2010. Neoplasia12, 39.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jogender Mehla et al.
Acta neuropathologica communications, 9(1), 193-193 (2021-12-17)
Previous reports indicate a potential role for signal transducer and activator of transcription 3 (STAT3) in amyloid-β (Aβ) processing and neuritic plaque pathogenesis. In the present study, the impact of STAT3 inhibition on cognition, cerebrovascular function, amyloid pathology, oxidative stress
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service