An ultrasensitive Coomassie-based reagent for staining polyacrylamide gels. No mixing or preparation is involved, no fixation step is needed, and no destaining is required. RAPIDstain Reagent stains only protein, within 5-10 min, and leaves a crystal-clear background with high resolution. Contain the following component:
• RAPIDstain 1 x 1 liter Suitable for 20 mini gels (8 x 10 cm) or 10 larger gels (12 x 15 cm)
An ultrasensitive Coomassie-based reagent for staining polyacrylamide gels. No mixing or preparation is involved, no fixation step is needed, and no destaining is required. RAPIDstain Reagent stains only protein, within 5-10 min, and leaves a crystal-clear background with high resolution. Note: 1 liter is sufficient to stain up to 20 minigels or 10 standard (16 x 18 cm) gels.
Warning
Toxicity: Harmful (C)
Other Notes
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany