Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

492029

Sigma-Aldrich

NRG1-β1/HRG1-β1 EGF Domain, Human, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human neuregulin 1-β1/heregulin 1-β1 epidermal growth factor-like domain (NRG1-β1/HRG1-β1 EGF Domain), consisting of amino acids 176-246 and expressed in E. coli. All NRG1 isoforms contain an EGF-like domain that is required for binding directly to the ErbB3 or ErbB4 receptor tyrosine kinases.

Biochem/physiol Actions

ED₅₀ = 0.06-0.3 ng/ml as measured in a serum-free proliferation assay using MCF-7 human breast adenocarcinoma cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered acetonitrile, TFA, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml with sterile PBS containing ≥1% human or bovine serum albumin.

Other Notes

Buonanno, A., and Fischbach, G.D. 2001. Curr. Opin. Neurobiol.11, 287.
Adlkofer, K. and Lai, C. 2000. Glia29, 104.
Garratt, A.N. et al. 2000. BioEssays22, 987.
Karey, K.P., et al. 1988. Cancer Res.48, 4083.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service