Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239783

Sigma-Aldrich

Cyclooxygenase Inhibitor Set

The Cyclooxygenase Inhibitor Set controls the biological activity of Cyclooxygenase. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

ambient

storage temp.

−20°C

General description

Contains 100 mg of Meloxicam (Cat. No. 444800), 5 mg of NS-398 (Cat. No. 349254), 5 mg of SC-560 (Cat. No. 565610), and 5 mg of Sulindac Sulfide (Cat. No. 574102).
Cyclooxygenases (COX) act on arachidonic acid to generate prostaglandins. They exist either as COX-1 or COX-2. COX-1, a constitutive enzyme associated with the endoplasmic reticulum, is responsible for maintaining normal physiologic function. It is found throughout the body, especially in the GI tract, kidneys, and platelets and its inhibition by NSAID is reported to cause GI damage, renal dysfunction, and platelet abnormalities. COX-2 is an inducible enzyme that is mainly associated with the nuclear envelope. It′s activity is induced by several growth factors, cytokines, and inflammatory signals. COX-2 expression is markedly increased in 85-90% of human colorectal adenocarcinomas, whereas COX-1 levels remain unchanged. Based on structural differences in the active sites of these isozymes, a new drug has been developed that specifically inhibits only the COX-2 activity. COX-2 inhibitors have the potential to provide the traditional benefits of NSAID with significantly reduced incidence of endoscopic ulcers. Hence, they offer greater therapeutic promise in the treatment of inflammation and cancer. This inhibitor set provides 4 inhibitors for testing the inhibitory effects on COX-1 and COX-2.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
COX-1
Secondary Target
COX-2

Packaging

Packaged under inert gas

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Fournier, D.B., et al. 2000. J. Cell Biochem.77, 97.
Crofford, L.J., et al. 2000. Arthritis Rheum.43, 4.
Masferrer, J.L., et al. 1999. Ann. N. Y. Acad Sci.889, 84.
Williams, C., et al. 1999. Ann. N. Y. Acad. Sci.889, 72.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service