Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-765-M

Sigma-Aldrich

BTK (R28H) Protein, active, 250 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

description

protein target BTK

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... BTK(695)

General description

N-terminal His6-tagged, recombinant, full-length, human BTK

Packaging

Also available in 10 μg size (cat#14-765) and in bulk (cat# 14-765-K).

Quality

routinely evaluated by phosphorylation of Src substrate peptide

Physical properties

78.4kDa

Storage and Stability

"Store -70°C. For maximum recovery of product, centrifuge original vial prior to removing the cap. Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C."

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service