MilliporeSigma
All Photos(4)
Key Documents
9502S-PP-CLR-K
MicroSolv
MicroSolv Polypropylene Vials
2 ml Vials, screw top, plastic. Translucent, 9-425 mm threads, 12x32 mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 1000/CS.
Synonym(s):
Non-ionic
About This Item
PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?
O.D.
12x32 mm
volume
2000 μL
2 mL
General description
Our most popular plastic autosampler vial can used instead of glass vials for HPLC. These high volume (2ml) blow-molded, plastic vials are made with virgin polypropylene, have fill lines with graduations and a slightly concave bottom that is helpful in reducing residual volume.
Non-ionic, these PP vials can reduce or eliminate adsorption of many compounds including proteins, peptides and bases where adsorption is common with borosilicate glass.
Excellent choice for Ion Chromatography, CE, Bio-analysis, analysis of PFAS or simply replacing basic HPLC autosampler vials. Compatible with most modern autosamplers that accept 12x32mm vials.