LIFH5F6BEB2B
Life Chemicals Inc.
2,3-diiodo-4,6-dimethylpyridine
Select a Size
Vendor SKU: F1957-0628-250MG
$284.70
Select a Size
About This Item
Vendor SKU: F1957-0628-250MG
$284.70
Iupac Name
2,3-diiodo-4,6-dimethylpyridine
Inchi Code
1S/C7H7I2N/c1-4-3-5(2)10-7(9)6(4)8/h3H,1-2H3
Inchi Key
ASHZLQUGKAWALO-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service