Documents
332-D419-OEM
Foxx Life Sciences
Foxx Life Sciences EZwaste® HPLC Solvent Waste System, Round HDPE Carboy Assembly,
HPLC Solvent Waste Assembly System, 20L RND Carboy, HDPE, 83mm Cap, 4x 1/8", 3x 1/4" OD Tubing, 1/4" & 3/8" HB & Carbon Filter, 1/EA
About This Item
PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Foxx Life Sciences
General description
Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter that traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste system also features a closed-loop design that protects against leaks and spills, improving overall lab safety.
The EZwaste® Solvent Waste System is designed to be an affordable solution for liquid waste disposal. Each system is manufactured to safely collect waste from one or more HPLC systems while the carbon exhaust filters improve air quality by capturing potentially harmful vapors emitted from solvents.
* Depending on the application, carbon exhaust filter life cycle may vary
  • Tube fittings secure waste tubes to carboy providing the closed system necessary to prevent spills (plugs included for unused ports)
  • 90° adapter included for maximum storage flexibility
  • Replaceable sulfur cured EPDM gasket for high chemical compatibility against aggressive organic solvents
  • Made to Order