Skip to Content
MilliporeSigma
All Photos(1)

Documents

ENAH03855FF6

Enamine

3-(trifluoromethyl)oxetan-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
142.08
MDL number:

Iupac Name

3-(trifluoromethyl)oxetan-3-ol

Inchi Code

1S/C4H5F3O2/c5-4(6,7)3(8)1-9-2-3/h8H,1-2H2

InChI key

DFMANELZPPUNRX-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Product Link

https://www.enaminestore.com/catalog/EN300-117987

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-117987


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service