CO1H324A5F62
CombiPhos Catalysts Inc
(6-isobutylpyridin-3-yl)boronic acid
Sign Into View Organizational & Contract Pricing
Select a Size
100 MG
$625.00
250 MG
$1,247.50
1 G
$1,875.00
5 G
$5,625.00
Vendor SKU: BA6079-100MG
$625.00
Ships in 30 days - from Aldrich Partner.
All Photos(1)
Select a Size
Change View
100 MG
$625.00
250 MG
$1,247.50
1 G
$1,875.00
5 G
$5,625.00
About This Item
CAS Number:
Molecular Weight:
179.03
Vendor SKU: BA6079-100MG
$625.00
Ships in 30 days - from Aldrich Partner.
Iupac Name
6-isobutyl-3-pyridinylboronic acid
Inchi Code
1S/C9H14BNO2/c1-7(2)5-9-4-3-8(6-11-9)10(12)13/h3-4,6-7,12-13H,5H2,1-2H3
Inchi Key
ATRWMVRMWAZAOO-UHFFFAOYSA-N
Purity
95%
Country of Origin
US
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service