Skip to Content
MilliporeSigma
All Photos(1)

Documents

CIAH991BE016

ChemScene LLC (Preferred Partner)

1-Cyclohexylpentane-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
182.26
MDL number:

Iupac Name

1-cyclohexyl-1,3-pentanedione

Inchi Code

1S/C11H18O2/c1-2-10(12)8-11(13)9-6-4-3-5-7-9/h9H,2-8H2,1H3

InChI key

BXPLVRDZCIJGLU-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/25790-34-5.html

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0243285.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service