Skip to Content
MilliporeSigma
All Photos(1)

Documents

CIAH987ED9E7

ChemScene LLC (Preferred Partner)

2-Bromo-5-(trimethylsilyl)-thiophene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
235.22
MDL number:

bp

70 C at 2 mmHg

Iupac Name

(5-bromothiophen-2-yl)trimethylsilane

Inchi Code

1S/C7H11BrSSi/c1-10(2,3)7-5-4-6(8)9-7/h4-5H,1-3H3

InChI key

IXLYRDOGOXBECE-UHFFFAOYSA-N

storage temp.

4C, stored under nitrogen

Purity

96%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/18246-28-1.html

Boiling point

70 C at 2 mmHg

Signal Word

Warning

Hazard Statements


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service